Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiocyanate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

Catalog No. LC249702
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC249702 1 L
LC249701 500 mL
LC249704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC249702 Supplier LabChem, Inc. Supplier No. LC249702
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-72-7 , 7732-18-5
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Color Colorless
CAS Max % 99.2, 0.8
Physical Form Liquid
Quantity 1 L
Linear Formula NaSCN
Solubility Information Soluble in water
Formula Weight 81.07
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000N ±0.0005N (0.1M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1g/mL
Chemical Name or Material Sodium Thiocyanate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.