Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiocyanate, Solstice

Catalog No. 6002228 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-28 100 g
60-022-30 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-022-28 Supplier Solstice Supplier No. 251410100G

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium;thiocyanate
SMILES [Na+].[S-]C#N

Specifications

Quantity 100 g
Linear Formula NaSCN
UN Number NONH for all modes of transport
Beilstein 3594965
Solubility Information Soluble in water, 1,000 g/L at 20°C
Formula Weight 81.07g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Sodium Thiocyanate
Hazard Statement P273-P280-P305 + P351 + P338
Precautionary Statement H302 + H312 + H332-H319-H412
EINECSNumber 208-754-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.