Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.01 N Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18613917 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-917 4 L Amber Glass Bottle
18-613-915 500 mL Amber Glass Bottle
18-613-916 1 L Amber Glass Bottle
18-613-918 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-613-917 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S5274LTGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-72-7 , 7732-18-5 , 10102-17-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Quantity 4 L
Concentration or Composition (by Analyte or Components) 0.01 N ± 0.0002 N
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.