Learn More
Specifications
Specifications
| Color | Colorless |
| Assay Percent Range | 0.02500 ± 0.00001 N at 20°C |
| Solubility Information | Solubility in Water: Infinite |
| SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
| IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
| Molecular Weight (g/mol) | 248.172 |
| Grade | Laboratory |
| Odor | Odorless to slight solvent odor |
| Packaging | Cubitainer |
| Solvent or Matrix | Aqueous Solution |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
