Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.500 N (N/2), Ricca Chemical
SDP

Catalog No. 796625 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
796625 10 L Cubitainer
7966-16 500 mL Natural Poly Bottle
7966-32 1 L Natural Poly Bottle
7966-1 4 L Natural Poly Bottle
79665 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 796625 Supplier Ricca Chemical Company Supplier No. 79662.5
Add to Cart
Add to Cart

Specifications

Color Colorless
CAS 10102-17-7, 7732-18-5, 497-19-8
CAS Min % 7.67, 90.0, 0.06
CAS Max % 7.98, 94.0, 0.06
Molecular Formula H10Na2O8S2
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17
PubChem CID 61475
ChEBI CHEBI:32150
Grade Laboratory
Packaging Cubitainer
pH ∼10
Quantity 10 L
Physical Form Liquid
Concentration or Composition Notes 0.500Normal
Chemical Name or Material Sodium Thiosulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.