Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, Pentahydrate, Crystal, BiotechGrade, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606884 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-884 2.5 kg Poly Bottle
18-606-882 500 g Poly Bottle
18-606-883 6 x 500 g Poly Bottle
18-606-885 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-884 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S14972.5KG
Add to Cart
Add to Cart

S1497, 10102-17-7, Na2S2O3·5H2O

Spectrum™ Chemical Sodium Thiosulfate, Pentahydrate, Crystal, BiotechGrade has been used to lessen some of the side effects of cisplatin (a cancer medicine). It is also used with another medicine in the emergency treatment of cyanide poisoning. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 100.5%
Grade Biotech
Odor Odorless
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.