Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, Pentahydrate, Crystal, Reagent, ACS, 99.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18606891 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-891 6 x 500 g Poly Bottle
18-606-892 2.5 kg Poly Bottle
18-606-893 4 x 2.5 kg Poly Bottle
18-606-894 12 kg Poly Pail
18-606-895 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-891 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1500500GMCS6
Add to Cart
Add to Cart

S1500, 10102-17-7, Na2S2O3·5H2O

Spectrum™ Chemical Sodium Thiosulfate, Pentahydrate, Crystal, Reagent, ACS - In analytical chemistry, the most important use comes because the thiosulfate anion reacts stoichiometrically with iodine in aqueous solutions. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

pH 6.0 to 8.4
CAS Max % 1
Quantity 6 x 500 g
Percent Purity 99.5 to 101%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.