Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Solution, SPEX CertiPrep™
SDP

Catalog No. 182002 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 gal.
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-200-2 1 gal. Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-200-2 Supplier SPEX CertiPrep Supplier No. 182002

missing translation for 'validMainframeMsg'

Prepared using double-deionized, ASTM Type I Water and packaged in a cubitainer to maximize matrix shelf life. SPEX CertiPrep™ Sodium Thiosulfate solutions are prepared from ACS Grade, micro-crystalline Na2S2O3·5H2O.

  • Ideal for ASTM D1510 applications

Chemical Identifiers

CAS 71-41-0 , 7732-18-5
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Physical Form Liquid
Quantity 1 gal.
Assay Percent Range 100%
Formula Weight 158.11 g/mol
Concentration or Composition (by Analyte or Components) 0.0394 N
Packaging Cubitainer
Chemical Name or Material Amyl Alcohol, H2O
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.