Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium trans-indole-3-acrylate, 96%, Thermo Scientific™

Catalog No. AAL1835103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1835103 1 g
AAL1835106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1835103 Supplier Thermo Scientific Chemicals Supplier No. L1835103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13615-46-8
Molecular Formula C11H8NNaO2
Molecular Weight (g/mol) 209.18
MDL Number MFCD02094005
InChI Key RVUGOFHDOUJAQL-UHFFFAOYSA-M
Synonym sodium 3-indoleacrylate, sodium 3-1h-indol-3-yl prop-2-enoate, 2-propenoic acid,3-1h-indol-3-yl-, sodium salt 1:1
PubChem CID 24212065
IUPAC Name sodium 3-(1H-indol-3-yl)prop-2-enoate
SMILES [Na+].[O-]C(=O)C=CC1=CNC2=CC=CC=C12

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 209.18
Percent Purity 96%
Chemical Name or Material Sodium trans-indole-3-acrylate
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.