Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium trifluoromethanesulfonate

Catalog No. p-7049744
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2083704 2 g
AA2083709 10 g
AA2083718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA2083704 Supplier Thermo Scientific Chemicals Supplier No. 02083704
Only null left
Add to Cart

CAS: 2926-30-9 | CF3NaO3S | 172.053 g/mol

Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2926-30-9
Molecular Formula CF3NaO3S
Molecular Weight (g/mol) 172.053
MDL Number MFCD00061607
InChI Key XGPOMXSYOKFBHS-UHFFFAOYSA-M
Synonym sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d
PubChem CID 23676748
IUPAC Name sodium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 253°C to 255°C
Color White
Odor Odorless
Linear Formula CF3SO3Na
Quantity 2 g
Beilstein 3728797
Sensitivity Hygroscopic
Solubility Information Soluble in water (1000g/L).
Formula Weight 172.06
Physical Form Powder
Chemical Name or Material Sodium trifluoromethanesulfonate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.