Learn More
Sigma Aldrich Fine Chemicals Biosciences Petroleum ether puriss. p.
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 322992.5L
Encompass
Petroleum ether is a low boiling point fraction having boiling point in the range of 40-60C. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory analgesic and antipyretic activities.
Catalog No. 50-289-8661
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.