Learn More
Sigma Organic Chemistry Sodium undecylenate | 250mg | 3398-33-2 | MFCD00054419
Supplier: Sigma Organic Chemistry CDS003850250MG

Sodium undecylenate , 250mg
About this Item:
CAS #: 3398-33-2
MDL #: MFCD00054419
Molecular Weight: 206.26
UNSPSC Code: 12352200
Chemical Formula: C11H19NaO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.