Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Soft Scrubs™ Short Sleeve Shirts

Catalog No. 19472448 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-448 X-Large
19-472-447 Small
19-472-446 Medium
19-472-451 Large
19-472-443 2X-Large
19-472-444 3X-Large
19-472-445 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-472-448 Supplier International Enviroguard™ Supplier No. 2055BXL
Add to Cart
Add to Cart

Disposable short sleeve SMS shirts are comfortable to wear for long durations and are ideal for controlled environments.

  • Short sleeve SMS shirts offer comfort and safety to work long hours
  • Offer modesty with complete ''no show through'' coverage
  • Can easily be worn as a protective undergarment beneath a sterile gown
  • After use, quickly and easily dispose to reduce spread of germs, bioburdens, and other contagions
  • Round hemmed neck comfortably lays flat
  • Left chest pocket allows storage for eyeglasses, small instruments, and pens
  • Applications include pharmaceuticals, labs, healthcare, controlled environments, and more
  • Anti-static fabric is made of an opaque material that doesn't show through and resists abrasions
  • Convenient, fabric loop for easily hanging up clothing
  • Elastic wrists stretch to provide snug, secure fit

Specifications

Size X-Large
Gender Unisex
Material SMS
Color Blue
Neck Style Hemmed Crew
Seam Type Serged
Sleeve Style Short Sleeve
Disposable Single-use
Latex Free Yes
No. of Pockets 2
Product Type Scrub Shirt
Sterility Non-sterile
For Use With (Application) Pharmaceutical, Lab, Medical, Manufacturing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.