Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ SoftCIDE™ Extra-Mild Antimicrobial Handwash

Catalog No. 1437925 Shop All Epredia Products
Change view
Click to view available options
Volume (English):
4 oz.
16 oz.
27 oz.
32 oz.
1 gal.
Packaging:
Bag-in-a-box carton refill
Pump Bottle
Refill Jug
Squeeze Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Packaging
14-379-25 27 oz. Bag-in-a-box carton refill
14-379-24 16 oz. Pump Bottle
12-544-09 32 oz. Pump Bottle
12-544-22 1 gal. Refill Jug
14-379-23 4 oz. Squeeze Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-379-25 Supplier Epredia™ Supplier No. 2102712001
This item is not returnable. View return policy

Effectively eliminate bacteria while replenishing the natural oils in your skin with Epredia™ SoftCIDE™ Extra-Mild Antimicrobial Handwash.

  • Uses 1% PCMX as an active ingredient
  • Unique formulation contains emollients
  • Effective against a broad spectrum or microorganisms
  • Soap is extremely gentle; pH balanced with the skin

Specifications

Product Type Extra Mild Anti-microbial Hand Wash
Description Bag-in-a-box carton refill
Packaging Bag-in-a-box carton refill
Volume (English) 27 oz.
Volume (Metric) 798 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.