Learn More
Thermo Scientific™ SoftFit-L™ Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Shop All Thermo Scientific Products

Description
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package.
- Hinged rack provides easy one-hand access to tips
- Color-coded inserts allow quick identification of tip volume
- Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin
Recommended for:
- Liquid Handling
- Molecular Biology
- PCR
- Biotechnology
- Front latch closes and locks, ensuring safe and secure storage throughout
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
- Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Lid rim prevents racks from sliding around when stacked and carted around a lab
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Not Autoclavable |
| Model | 1000 μL, Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit |
| Length (English) | 3.39 in. |
| Length (Metric) | 8.61 cm |
| Certifications/Compliance | DNase, RNase, DNA, ATP, and Endotoxin Free |
| Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.