Learn More
PIP™ SoftStar™ Disposable Soft Polyurethane Foam Corded Ear Plugs - NRR 30
Catalog No. 17489665 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
30
Catalog No. Noise Reduction Rating (NRR)
17-489-665 30
1 options
Catalog No. 17-489-665 Supplier PIP™ Supplier No. 267HPF710C
Add to Cart
Add to Cart
Disposable soft polyurethane foam corded ear plugs - NRR 30.
  • Innovative design in a traditional cylindrical shape
  • The smoothest surface or 'skin' of any cylindrical shaped ear plug
  • Unlike other ear plugs, the SoftStar Cylinder has a foam handle for ease of removal
  • Black PVC cord for convenience and to prevent loss of ear plugs
  • NRR 30 protection
Specifications
Size Universal
Noise Reduction Rating (NRR) 30
Corded Corded
Plug Shape Cylinder
Product Type Disposable Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric Yes