Learn More
Syngene™ Dymension 1 Software
GREENER_CHOICESDP
Catalog No. 01257181 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Gbox-Chemi-XX6/9
Catalog No. For Use With (Equipment)
01-257-181 Gbox-Chemi-XX6/9
1 options
Catalog No. 01-257-181 Supplier Syngene™ Supplier No. DYM1
Add to Cart
Add to Cart
Syngene™ Dymension 1 Software is a 2D analysis software for gel electrophoresis.
  • Software for the rapid analysis of 2D protein spots
    between samples containing one gel only
  • Synchronised zoom with intensity autoscale
  • Automatic and precise image warping with vector view
  • Background correction and noise filtering
  • Automatic spot detection 3 x 10MB images
  • Spot detected in eight seconds
  • Automatic spot matching
  • Spot detection with user adjustable parameters
  • Spot editing and filtering Spot normalization
Specifications
For Use With (Application) 2d electrophoresis analysis
For Use With (Equipment) Gbox-Chemi-XX6/9