Learn More
Syngene™ Dymension 3 Software
GREENER_CHOICESDP
Catalog No. 01257183 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Gbox-Chemi-XX6/9
Catalog No. For Use With (Equipment)
01-257-183 Gbox-Chemi-XX6/9
1 options
Catalog No. 01-257-183 Supplier Syngene™ Supplier No. DYM3
Add to Cart
Add to Cart
Request a Quote
Syngene™ Dymension 3 Software is a 2D analysis software for gel electrophoresis.
  • Software for the rapid analysis of 2D protein spots from multiple gels including multi-stained fluorescent gels (Cy2, Cy3, Cy5)
  • With interactive process flow diagram
  • Main pane and secondary panes for viewing replicates and samples
  • Synchronised zoom with intensity autoscale
  • Automatic and precise image warping with vector view
  • Manual warp editing
  • Automatic background subtraction, noise filtering
  • SYPRO Ruby crystal filter
  • Automatic spot detection
  • Region of interest option
  • Gaussian spot modelling option
  • 3 x 10MB images spot detected in eight seconds
  • Spot detection with user adjustable parameters
  • Spot editing, filtering and normalisation
  • Spot matching across multiple gels
  • Manual match editing pI and MW ladder calibration
  • Protein list assignment of spots with known identity, pI and MW
  • Histogram (brightness and contrast)
  • Main pane and secondary panes for multi-stained gel sets, replicates and samples
  • Bar chart 3D spot profile
  • Matched spot plot
  • Correlation plot
  • Table of spot matching results
  • Scatter plot
  • Statistical tests
  • Export to Word and Excel
  • Spot picker interface
  • Supplied with one years' support
Specifications
For Use With (Application) 2d electrophoresis analysis
For Use With (Equipment) Gbox-Chemi-XX6/9