Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOHLH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116291
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116291 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116291 Supplier Thermo Scientific Supplier No. PA5116291
Only null left
Add to Cart
Add to Cart

SOHLH2 Polyclonal Antibody

SOHLH2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SOHLH2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SOHLH2
Gene Accession No. Q9NX45
Gene Alias 4933406N12Rik; AI413378; bHLHe81; RGD1304752; Sohlh2; SPATA28; spermatogenesis and oogenesis specific basic helix-loop-helix 2; spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 2; spermatogenesis associated 28; TEB1; testicular secretory protein Li 50; testicular secretory protein Li 51; Tohlh2
Gene Symbols SOHLH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 232-258 amino acids from the Central region of human SOHLH2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.