Learn More
Cayman Chemical Solamargne 5mg
SDP
Supplier:  Cayman Chemical 281105 MG
Encompass_Preferred
A potent agonist of GPR35 (EC50 1 61 nM) inhibits histamine release in isolated rat peritoneal mast cells (IC50s 0 1-50 UM) inhibits A23187-induced calcium influx in isolated rat mast cells reduces antigen-induced histamine release from rat conjunctival tissue by 46 in vitro when used at a concentration of 10 Ug/ml dose-dependently reduces the immediate hypersensitivity response in rat conjunctiva in vivo reduces mast cell degranulation in rat conjunctiva
Catalog No. 50-293-7135
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.