Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana Instrument
GSA_VASDP

Catalog No. 23043111 Shop All QuidelOrtho Products
Change view
Click to view available options
Depth (English):
9.4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
23-043-111 9.4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-111 Supplier Quidel Supplier No. M900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

1 Purchased Solana™, Instrument, 95C Lysis Heat Block, Printer, Labels, Tray, 3 Racks and Stylus

Bench top instrument that combines Quidel’s proprietary helicase-dependent amplification (HDA) with fluorescence detection to deliver molecular results you can trust, faster than ever before.

Isothermal amplification

  • Ensures molecular sensitivity with moderate complexity
  • Eliminates thermal cycling to aid in faster time to result (as little as 30 minutes)

Simplified workflow

  • Avoids the need for specialized molecular operator training, making Solana easily adoptable

    Small foot-print

  • Facilitates decentralization of the instruments to bring them closer to the patient

Data Management

Results can be reported and stored in multiple ways to meet your data management needs:

  • USB data export to assist in the transfer of data without the need for direct connection
  • Optional printing allows for storage of hardcopy results which may enhance post-analytical processes
  • Test, QC, and Calibration results stored on-board
  • LIS and Middleware capable
  • Available with the power of Virena

Security

Solana offers supervisors the ability to set access rights by operator to prevent unauthorized use or use by untrained operators

Barcode technology

Reduce time and errors associated with manually recording patient and operator ID

Connectivity

Reduce time manually recording test results and risk of transcription errors. Connect to many of existing information systems.

Molecular multi-assay platform with ability to run for

  • Group A Strep
  • Strep Complete (A, C, and G)
  • Influenza A+B
  • Trichomonas
  • HSV 1&2/VZV

Product Specifications:

  • Dimensions: 9.4 in deep x 9.4 in wide x 5.9 in high
  • Weight: 8.8 lb.
  • Power Supply Input: 100/240 VAC, 47/63 Hz, 1.5-0.7 Amps
  • Operating temperature: 15°C to 35°C/59°F to 86°F
  • Indoor use only
  • Elevation up to 2,000m high
  • Operating humidity: Highest relative humidity 80% for temperatures up to 31°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Depth (English) 9.4 in.
Height (English) 5.9 in.
Width (English) 9.4 in.
Weight (English) 8.8 lb.
Electrical Requirements 100/240 V 47/63 Hz 1.5-0.7 A
For Use With (Application) Group A Strep; Strep Complete (A, C, and G); Influenza A+B; Trichomonas; HSV 1 and 2/VZV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.