Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana SARS-CoV-2 LYO Assay
SDP

Catalog No. 23044178 Shop All QuidelOrtho Products
Change view
Click to view available options
Includes:
48 Tests Only
48 Tests, 50 UTM/NS swab
48 Tests, 50 UTP/NP swab
Starter Kit- 48 Tests, 50 UTM Swab/ Analyzer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-044-178 48 Tests, 50 UTP/NP swab
23-044-177 48 Tests Only
23-044-179 48 Tests, 50 UTM/NS swab
23-044-180 Starter Kit- 48 Tests, 50 UTM Swab/ Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-044-178 Supplier Quidel Supplier No. M960
Add to Cart
Add to Cart
This item is not returnable. View return policy

An isothermal Reverse Transcriptase - Helicase-Dependent Amplification (RT-HDA) assay intended for the qualitative detection of nucleic acid from SARS-CoV-2 in nasopharyngeal (NP) and nasal (NS) swab specimens from individuals suspected of COVID-19.

  • RT-HDA technology: Rapid method of isothermal nucleic acid amplification that does not require thermal cycler
  • Small, easy-to-use instrument
  • Room temperature set up
  • Long shelf life: 6 months from date of manufacture

Specifications

CE Marker CE-IVD
Clia Complexity Moderate
Detectable Analytes SARS-CoV-2
Format Tube
Disposable Single-use
For Use With (Application) RT-PCR, Rapid PCR
Includes 48 Tests, 50 UTP/NP swab
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.