Learn More
Description
- Molecular Biology grade. D
- pH of solution at 25°C: 6.85-7.15
- Protease: None detected
- RNase: None detected
- Rapid in vitro diagnostic test
- For qualitative detection of nucleic acids isolated from clinician-collected vaginal swab specimens or urine specimens obtained from asymptomatic or symptomatic patients
- Competitive process control (PRC) included Lysis Tube
- Monitor sample processing, inhibitory substances in clinical samples, reagent failure or instrument failure
- HDA Technology: Rapid method of nucleic acid amplification that does not require thermal cycler
- CLIA Moderate: Requires minimal training
- Lypholized Master Mix Reagent: Easy-to-use format, just add processed sample to rehydrate
- Refrigerated Storage: No freezer needed, 2 to 8˚C storage
- Room temperature setup: No ice or cooling block required
- Urine sample specimen
- Results are displayed on the touchscreen, can be saved to the instrument, printed
- Capable of being sent to LIS and exported through one of Solana’s five USB ports.
- Supported by the power of Virena™.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Assay |
| Clia Complexity | Moderate |
| Sample Type | Vaginal Swab |
| Sensitivity | 100% (Asymptomatic), 98.6% (Symptomatic), 99.2% (All) |
| Type | Assay |
| For Use With (Application) | For aid in diagnosis of Trichomonas |
| Includes | Test Kit: Package insert, Dilution Buffer, Lysis Buffer, Reaction Tubes; Lyophilized Master Mix reagent |
| Quantity | 48 Tests/Kit |
| Specificity | 98.9% (Asymptomatic)/ 98.5% (Symptomatic)/98.7% (All) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

