Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Solana Trichomonas Starter Kit
GSA_VASDP

Catalog No. 23043103 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Assay
Starter Kit
Sample Type:
Urine
Vaginal Swab
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Sample Type
23-043-103 Assay Urine
23-043-105 Starter Kit Vaginal Swab
23-043-104 Starter Kit Urine
23-043-102 Assay Vaginal Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-043-103 Supplier Quidel Supplier No. M304.U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Delivers molecular results faster with bench top testing system. Quidel™ Solana™ Instrument with Trichomonas Urine Test Kit uses helicase-dependent amplification (HDA) with fluorescence detection

  • Molecular Biology grade. D
  • pH of solution at 25°C: 6.85-7.15
  • Protease: None detected
  • RNase: None detected

Assay Features:
  • Rapid in vitro diagnostic test
  • For qualitative detection of nucleic acids isolated from clinician-collected vaginal swab specimens or urine specimens obtained from asymptomatic or symptomatic patients 
  • Aids in the diagnosis of Trichomoniasis.
  • Competitive process control (PRC)
  • Monitors sample processing, inhibitory substances in clinical samples, reagent failure or instrument failure
  • HDA Technology: Rapid method of nucleic acid amplification that does not require thermal cycler
  • CLIA Moderate: Requires minimal training
  • Lypholized Master Mix Reagent: Easy-to-use format, just add processed sample to rehydrate
  • Refrigerated Storage: No freezer needed, 2 to 8˚C storage
  • Room temperature setup: No ice or cooling block required
  • Urine sample specimen
 
Results Display
  • Touchscreen, can be saved to the instrument, printed
  • Capable of being sent to the LIS and exported through one of Solana’s five USB ports.
  • Supported by the power of Virena™.


  

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Description Assay
Clia Complexity Moderate
Sample Type Urine
Sensitivity 98.0% (Asymptomatic), 92.9% (Symptomatic), 95.0% (All)
Type Assay
For Use With (Application) For aid in diagnosis of Trichomonas
Includes Test Kit: Package insert, Dilution Buffer, Lysis Buffer, Reaction Tubes; Lyophilized Master Mix reagent
Quantity 48 Tests/Kit
Specificity 98.4% (Asymptomatic)/ 97.9% (Symptomatic)/98.2% (All)
Test Time ≈35 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.