Learn More
Solar-Cult™ Bacteria/Fungi Dipslides, AquaPhoenix Scientific™
SCI_ED
Catalog No. S88636 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
10 pk
Catalog No.
S88636 10 pk
1 options
Catalog No. S88636 Supplier Aqua Phoenix Scientific Supplier No. MC600
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Dip slides are a popular way to check microbial levels in cooling systems.
Specifications
Detects Bacteria/Fungi
Quantity 10 Pack