Learn More
United Scientific Supplies Solar Powered Car
SDPSCI_ED
Catalog No. S02601 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Activity guide
Catalog No. Includes
S02601 Activity guide
1 options
Catalog No. S02601 Supplier United Scientific Supplies Supplier No. SLRCR1
Add to Cart
Add to Cart
Economical and attractive demonstration Solar Powered Car model.
  • Powered by a solar cell or a rechargeable battery.
  • Battery can be recharged from the solar cell, demonstrating an electricity storage system for solar power.
  • Solar cell can be tilted to maximize the solar input.
  • Solar cell output can be measured at two binding posts.
  • Slide switch turns on/off and selects function.
  • All working parts visible through clear molded housing.
Order Info
Dimensions: 5.75 x 2.75 x 3 in. (14.5 x 6.9 x 8.8cm). Weights: Product: 4.5 oz (129g) Packed item: 6.5 oz (221g). Product with battery: 5.5 oz (157g)
Specifications
Product Type Solar Car
Includes Activity guide