Learn More
Solid Glass Beads
SCI_ED
Increase the efficiency of separations.
Specifications
Material Glass
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity & Availability  
S800242
SCI_ED
View Documents
Walter Stern Inc.
100F/EMD
5 mm 2700/Pk. N/A N/A N/A
S80024
SCI_ED
View Documents
Walter Stern Inc.
100C/EMD
3 mm 12500/Pk. N/A N/A N/A
S800241
SCI_ED
View Documents
Walter Stern Inc.
100E/EMD
4 mm 5500/Pk. N/A N/A N/A
S800243
SCI_ED
View Documents
Walter Stern Inc.
100G/EMD
6 mm 1600/Pk. N/A N/A N/A
Description
Description
  • For use in distillations or as inert boiling stones
  • Packaged in 1 lb. (0.45kg) packs
Specifications
Specifications
Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.