Learn More
Sigma Aldrich Fine Chemicals Biosciences Solid glass beads borosili
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z273619
Encompass
For use as distillation column packing mixing beads and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. /-10% variation in bead size.
Catalog No. 50-290-8768
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.