Learn More
United Scientific™ Solid Glass Stoppers
SDPSCI_ED
Catalog No. S012416 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Standard Taper Joint:
#13
#16
#19
#22
#27
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
S012416 #19
S012418 #13
S012417 #16
S012415 #22
S012414 #27
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S012416 Supplier United Scientific™ Supplier No. UNSGS19
Add to Cart
Add to Cart
Easily seal ground jointed glassware like flasks and separatory funnels with these Solid Glass Stoppers, featuring interchangeable ground joints.
  • Solid penny head glass stoppers for use with ground jointed glassware, such as flasks and separatory funnels
  • For ASTM standard flasks
  • Made of borosilicate 3.3 glass
Specifications
Standard Taper Joint #19
Material Borosilicate Glass
Shape Tapered
Stopper Number 19
Product Type Solid Pennyhead Stopper
Length (Metric) 46 mm
Autoclavable Autoclavable
Color Clear
Diameter (Metric) Outer 18 mm