Learn More
Synthware™ Solid Penny-Head Stopper
GREENER_CHOICESDP
Catalog No. 31501879 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
10/18
10/30
14/20
19/22
24/40
29/42
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-501-879 19/22
31-501-876 10/18
31-501-877 10/30
31-501-878 14/20
31-501-880 24/40
31-501-881 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-501-879 Supplier Synthware™ Supplier No. S291922
Add to Cart
Add to Cart
Flask length solid penny head stoppers
Specifications
Quantity 1 Ea.
Standard Taper Joint 19/22
Color Clear
Material Borosilicate Glass