Learn More
Solketal, 97%
Description
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .
Solubility
Soluble in water (fully), alcohol, acetals, esters, and ethers.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are acids, Strong oxidizing agents.
Specifications
Specifications
| Melting Point | -27°C |
| Density | 1.064 |
| Boiling Point | 188°C to 190°C |
| Flash Point | 80°C (176°F) |
| Refractive Index | 1.434 |
| Quantity | 2500 g |
| Beilstein | 104465 |
| Merck Index | 14,5213 |
| Solubility Information | Soluble in water (fully),alcohol,acetals,esters,and ethers. |
| Formula Weight | 132.16 |
| Show More |
Safety and Handling
| Hazard Category | H227-H319 |
| Precautionary Statement | P210-P264b-P280-P305+P351+P338-P370+P378q-P501c |
| EINECSNumber | 202-888-7 |
| RTECSNumber | JI0400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.