Learn More
Creative Biomart BNT162a1 SARS-CoV-2 Vaccine Antigen
Supplier:  Creative Biomart THP0325
Encompass_Preferred
BNT162a1 is one of four advanced mRNA-based vaccines being developed through "Project Lightspeed," a joint program between Pfizer and BioNTech. BNT162a1 is a uridine mRNA (uRNA) vaccine formulated in lipid nanoparticles for delivery via intramuscular injection.BNT162a1 is undergoing evaluation in clinical trials in both the USA (NCT04368728) and Germany (NCT04380701).
Catalog No. 50-208-4838
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.