Learn More
3M™ Solus™ 1000 Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Catalog No. 19169042 Shop All 3M Products
Change view
Click to view available options
Product Type:
Safety Glasses
Safety Glasses Kit
Frame Color:
Blue, Black
Green, Black
Lens Tint:
Amber
Clear
Gray
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Frame Color Lens Tint
19-169-042 Safety Glasses Green, Black Amber
19-169-044 Safety Glasses Blue, Black Clear
19-169-047 Safety Glasses Kit Blue, Black Clear
19-169-045 Safety Glasses Blue, Black Gray
19-169-046 Safety Glasses Blue, Black Amber
19-169-040 Safety Glasses Green, Black Clear
19-169-043 Safety Glasses Kit Green, Black Clear
19-169-041 Safety Glasses Green, Black Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-169-042 Supplier 3M™ Supplier No. 7100079181
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
3M™ Solus™ Safety Glasses, feature a clear, strong polycarbonate lens in a slim, stylish, green and black-accented colour frame. Our safety glasses have 3M™ Scotchgard™ Anti-Fog / Anti-Scratch Coating, and a soft temple and nose bridge.
  • Stylish and modern look slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
  • Lightweight with soft cushioning on nose bridge
  • Inner padding on temples for increased comfort and stability when worn
  • Meets K&N marking requirements of EN 166
  • Frame marking: 3M EN166 FT CE
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1+ 2015, CSA Z94.3
Lens Coating Anti-fog
Lens Tint Amber
Product Type Safety Glasses
For Use With (Application) Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation
Frame Color Green, Black
Model S1203SGAF
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Half Frame