Learn More
3M™ Solus™ 1000 Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Catalog No. 19169043 Shop All 3M Products
Change view
Click to view available options
Product Type:
Safety Glasses
Safety Glasses Kit
Frame Color:
Blue, Black
Green, Black
Lens Tint:
Amber
Clear
Gray
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Frame Color Lens Tint
19-169-043 Safety Glasses Kit Green, Black Clear
19-169-044 Safety Glasses Blue, Black Clear
19-169-047 Safety Glasses Kit Blue, Black Clear
19-169-045 Safety Glasses Blue, Black Gray
19-169-046 Safety Glasses Blue, Black Amber
19-169-040 Safety Glasses Green, Black Clear
19-169-041 Safety Glasses Green, Black Gray
19-169-042 Safety Glasses Green, Black Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-169-043 Supplier 3M™ Supplier No. 7100079182
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Solus™ Safety Glasses, feature a clear, strong polycarbonate lens in a slim, stylish, green and black-accented colour frame. Our safety glasses have 3M™ Scotchgard™ Anti-Fog / Anti-Scratch Coating, and a soft temple and nose bridge.
  • Stylish and modern look slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
  • Lightweight with soft cushioning on nose bridge
  • Inner padding on temples for increased comfort and stability when worn
  • Meets K&N marking requirements of EN 166
  • Frame marking: 3M EN166 FT CE
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1+ 2015
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Glasses Kit
For Use With (Application) Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation
Frame Color Green, Black
Includes Removable foam gasket and elastic strap
Model S1201SGAF-KT
Temple Style Removable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Half Frame