Learn More
3M™ Solus™ 1000 Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
3M™ Solus™ Safety Glasses, feature a clear, strong polycarbonate lens in a slim, stylish, green and black-accented colour frame. Our safety glasses have 3M™ Scotchgard™ Anti-Fog / Anti-Scratch Coating, and a soft temple and nose bridge.
Supplier:  3M™ 7100079184
 View more versions of this product
Catalog No. 19-169-045
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Stylish and modern look slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
  • Lightweight with soft cushioning on nose bridge
  • Inner padding on temples for increased comfort and stability when worn
  • Meets K&N marking requirements of EN 166
  • Frame marking: 3M EN166 FT CE
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ANSI Z87.1+ 2015, CSA Z94.3
Gray
Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation
S1102SGAF
Polycarbonate
Half Frame
Anti-fog
Safety Glasses
Blue, Black
Removable
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.