Learn More
3M™ Solus™ 2000 Series Prescription Safety Glasses
Catalog No. 19072205 Shop All 3M Products
Change view
Click to view available options
Description:
Reading Diopter: +1.5
Reading Diopter: +2.0
Reading Diopter: +2.5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-072-205 Reading Diopter: +2.5
19-072-204 Reading Diopter: +1.5
19-072-206 Reading Diopter: +2.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-072-205 Supplier 3M™ Supplier No. 7100222135
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Solus™ 2000 Series Prescription Safety Glasses provide eye protection with a stylish flare
  • Adjustable ratchet temples allow the wearer to customize their fit
  • Readers with anti-fog and anti-scratch coating that come in a range of diopters: +1.5, +2.0, or +2.5.
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays
  • Meet the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Description Reading Diopter: +2.5
Lens Tint Clear
Product Type Model: S2025AF-BLU
For Use With (Application) Machine Operations, Sub Applications, Law Enforcement, Power Fastening, Chemical Handling, Mining, Pouring and Casting, Painting, General Manufacturing, Machining, Assembly, Food and Beverage Manufacturing, Oil and Gas, Primary Metals, Pharmaceuticals, Chiseling, Grinding, Chemicals, Drilling, Aviation, Facility Maintenance, Automotive, Demolition, Masonary, Food and Beverage, Remediation, Construction
Frame Color Gray
No. per Case 20/Cs.
Temple Style Dual Injected Temple
Temple Color Blue, Gray
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Half Frame