Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Solution dNTPs: PCR Nucleotide Mix

Catalog No. 45001459 Shop All Cytiva Products
Change view
Click to view available options
Concentration:
10 mM
25 mM
Quantity:
500 μL
500 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Quantity
45-001-459 25 mM 500 μm
45-001-461 10 mM 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-459 Supplier Cytiva Supplier No. 28406560
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Free from DNase, RNase, and nicking enzyme activity

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type PCR Nucleotide Mix dNTP Mix (25mM each A,C,G,T)
Concentration 25 mM
Purity >99%
Quantity 500 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.