Learn More
ENG Scientific Solution II - Acid Alcohol (3% HCL)
Solution II - Acid Alcohol (3% HCL) is ideal for staining acid-fast bacteria. Use with Acid-fast stain kit. Modification of the Kinyoun Method.
Supplier:  ENG Scientific 6510
 View more versions of this product
Catalog No. ES6510
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Acid-fast bacilli stain red; all other bacteria stain blue with Sol. III, green with Sol. IIIA.
  • Solution II - Acid Alcohol for Acid Fast Stain Kit (Modification of the Kinyoun Method)
  • Kit includes: Solution I - Carbol Fuchsin, Solution II - Acid Alcohol, Solution III - Loeffler's Methylene Blue and Solution IIIA - Brilliant Green
  • For in vitro diagnostic use
  • Uses certified dyes
  • Specifications
    Specifications
    Bottles
    2 x 250 mL
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Title
    Select an issue
    For in vitro diagnostic use only.