Learn More
ENG Scientific Solution II - Acid Alcohol (3% HCL)
Solution II - Acid Alcohol (3% HCL) is ideal for staining acid-fast bacteria. Use with Acid-fast stain kit. Modification of the Kinyoun Method.
$233.00 - $646.00
Specifications
Packaging Bottles
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ES6510
View Documents
ENG Scientific
6510
2 x 250 mL
Pack of 2 for $233.00
Only null left
Add to Cart
ES6512
View Documents
ENG Scientific
6512
1 gal.
Each for $646.00
Only null left
Add to Cart
Description
Description
  • Acid-fast bacilli stain red; all other bacteria stain blue with Sol. III, green with Sol. IIIA.
  • Solution II - Acid Alcohol for Acid Fast Stain Kit (Modification of the Kinyoun Method)
  • Kit includes: Solution I - Carbol Fuchsin, Solution II - Acid Alcohol, Solution III - Loeffler's Methylene Blue and Solution IIIA - Brilliant Green
  • For in vitro diagnostic use
  • Uses certified dyes
  • Specifications
    Specifications
    Bottles
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    For in vitro diagnostic use only.