Learn More
Glass Expansion Solva Flex Pump Tube 2tag, 140mm spacing, 0.76mm ID, Black/Black (PKT 12)
SDP
Supplier:  Glass Expansion 0.76BLKSF
Encompass_Preferred
Solva Flex Pump Tube 2tag, 140mm spacing, 0.76mm ID Black/Black (PKT 12).Tag Colors: Black/BlackID mm: 0.76Wall Thickness mm: 0.86OD mm: 2.48Overall Length: 455mm (140mm spacing between tags)Number of Tubes Per Pack: 12Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon 4040 or equivalent.Special Properties: Special tubing for hydrocarbons, petroleum products and distillates.Chemical Compatibility: Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.Limitations: Not recommended for strong acids and alkalis, foodstuffs, beverages and medicines. Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6818
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.