Learn More
Optimize Technologies OPTI-SOLV Solvent Reservoir Filter with Tube Stem for 1/8" ID Tubing, 2µm, 5pk
SDP
Supplier:  Optimize Technologies 100400119
Encompass_Preferred
OPTI-SOLV™ Reservoir Filters employ a unique conical design inside the filter housing to prevent air bubbles from getting trapped and disturbing your analysis. While the availability of HPLC-grade solvents diminishes the need for pre-filtering, it is still imperative that a filtration device be installed in the reservoir. Placing a filter in the reservoir is an economical way to filter particles that may result from buffer salt precipitation, airborne dust, improperly cleaned glassware, or microbial contamination; none of which are remedied by the use of HPLC-grade solvents. These specific filters are made of Hastelloy C™ offering maximum corrosion resistance and inertness and are recommended for systems exceeding flow rates of 10 mL/minute.
Catalog No. 50-113-267
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.