Learn More
Sigma Aldrich Fine Chemicals Biosciences Solvent SafeTM pipette tip
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S79391CS
Encompass
Solvent Safe carbon filter pipette tipsfilters block corrosive vapors created when pipetting volatile organic solvents such as TfASafely and accurately pipette phenol/chloroform DNA extractionsPerform peptide synthesis and HPLC analysisExcellent alternative to glass pipettes
Catalog No. 50-290-4226