Learn More
CPC Scientific H-Ser-Ala-Asn-Ser-Asn-Pro-Ala-Met-Ala-Pro-Arg-Glu-Arg-Lys-Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SOMA008B
Encompass
Peptide that arises from the posttranslational cleavage of prosomatostatin, which in turn is derived from a large precursor, preprosomatostatin. It was found in the central and peripheral nervous system, the gastrointestinal (GI) tract. It additionally increases amylase secretion, cellular cycle GMP, and Ca2+ outfluxes.SEQUENCE: H-Ser-Ala-Asn-Ser-Asn-Pro-Ala-Met-Ala-Pro-Arg-Glu-Arg-Lys-Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys-OH (trifluoroacetate salt)(Cys17 and 28 bridge)ONE-LETTER SEQUENCE: SANSNPAMAPRERKAGCKNFFWKTFTSC (Cys17 and 28 bridge)MOLECULAR FORMULA: C137H207N41O39S3MOLECULAR WEIGHT: 3148.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [73032-94-7]SYNONYMS: Somatostatin-28RESEARCH AREA: GastrointestinalREFERENCES:P. Bohlen et al., BBRC, 96, 725 (1980)L. Pradayrol, et al., FEBS Letters, 109, 55 (1980)SKU(s): SOMA-008A, SOMA-008B
Catalog No. 50-211-0434
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.