Description
- The green oxygen hose assembly is provided with standard medical DISS Female threaded fittings on both ends
- The DISS fitting mates with the DISS Male on the anesthesia system
- Medical grade DISS Oxygen fittings
Specifications
Specifications
| For Use With (Application) | Research Animal Inhalant Anesthesia |
| Length (English) | 6 ft. |