Learn More
SOMNI Scientific™ SOMNI Scientific™ O2 Hose Assembly DISS
SDP
Catalog No. 04777220 Shop All Somni Scientific Products
Change view
Click to view available options
Length (English):
3 ft.
6 ft.
9 ft.
12 ft.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
04-777-220 9 ft.
04-777-218 3 ft.
04-777-219 6 ft.
04-777-221 12 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 04-777-220 Supplier SOMNI Scientific™ Supplier No. HF10011
Add to Cart
Add to Cart
Provided with standard medical DISS Female threaded fittings on both ends The DISS fitting mates with the DISS Male on the anesthesia system
  • The green oxygen hose assembly is provided with standard medical DISS Female threaded fittings on both ends
  • The DISS fitting mates with the DISS Male on the anesthesia system
  • Medical grade DISS Oxygen fittings
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Length (English) 9 ft.