Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SON Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115947 Supplier Invitrogen™ Supplier No. PA5115947
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SON.

The protein encoded by this gene binds to a specific DNA sequence upstream of the upstream regulatory sequence of the core promoter and second enhancer of human hepatitis B virus (HBV). Through this binding, it represses HBV core promoter activity, transcription of HBV genes, and production of HBV virions. The protein shows sequence similarities with other DNA-binding structural proteins such as gallin, oncoproteins of the MYC family, and the oncoprotein MOS. It may also be involved in protecting cells from apoptosis and in pre-mRNA splicing. Several transcript variants encoding different isoforms have been described for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SON
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SON
Gene Accession No. P18583, Q9QX47
Gene Alias 2900011L12Rik; AA409051; AU067731; BASS1; Bax antagonist selected in Saccharomyces 1; C21orf50; C81487; DBP5; DBP-5; HSPC310; HSPC312; KIAA1019; mKIAA1019; negative regulatory element-binding protein; NRE-binding protein; NREBP; Protein DBP-5; Protein SON; SON; Son cell proliferation protein; Son DNA binding protein; SON DNA-binding protein; SON3
Gene Symbols SON
Host Species Rabbit
Immunogen A synthesized peptide derived from human SON(Accession P18583), corresponding to amino acid residues K1804-S1854.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20658, 304092, 6651
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.