Learn More
Enzo Life Sciences Sonidegib diphosphate (10mg). CAS: 1218778-77-8
SDP
Supplier:  Enzo Life Sciences ENZCHM2370010
Encompass_Preferred
Sonidegib, also known as LDE-225, is a potent and selective smoothened (Smo) antagonist. Alternative name: LDE225 . diphosphate. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3658
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.