Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ Thermosafe EPS Foam-Only Shipper Foam Rings

Catalog No. 03001107 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Size:
2 in. (5.08 cm) ring
3 in. (7.62 cm) ring
Wall Thickness (English):
1.5 in.
3 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Wall Thickness (English)
03-001-107 3 in. (7.62 cm) ring 1.5 in.
03-001-250 3 in. (7.62 cm) ring 3 in.
03-001-112 2 in. (5.08 cm) ring 1.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-001-107 Supplier Sonoco™ Supplier No. 808R3

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Increase the flexibility of your warehouse inventory by utilizing foam rings on standard shippers.

  • Patented components basic container (body and dome lid), and stackable rings enable you to just add rings to increase package height
  • Create a perfect match between payload and container capacity
  • Increase packing flexibility while decreasing packaging inventory
  • Simplify ordering warehouse only one box footprint plus rings to vary capacity as needed
  • Designed to maximize temperature control and withstand the hazards of transport
  • Available for eight of our most popular size foam containers
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 foam only shipper
Applications ThermoSafe™ EPS insulated shipper having 12.5 L x 12.5 W x 6 in. H inner and 15.5 L x 15.5 W x 9 in. H outer dimension
Size 3 in. (7.62 cm) ring
Type Foam Ring
Wall Thickness (English) 1.5 in.
Material Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.