Learn More
Sonoco™ ThermoSafe Insulated Foam Shipper
EPS foam-only shippers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen products
$161.00
Specifications
Type Insulated Shipper
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (LxWxH) Dimensions (L x W x H) Exterior Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (LxWxH) Dimensions (L x W x H) Exterior Quantity Price Quantity & Availability  
03-001-210
View Documents
Sonoco™
421
ID: 19.375 x 11.25 x 3.875 in. 58.4 x 40.6 x 53.3 cm 3/Cs.
Pack of 3 for $161.00
Only null left
Add to Cart
03-001-207
View Documents
Sonoco™
668
ID: 21.56 x 13.43 x 10.62 in. 68.5 x 66.0 x 45.7 cm 2/Cs. N/A N/A N/A
03-001-203
View Documents
Sonoco™
671
ID: 23.25 x 15.81 x 6.31 in. 68.5 x 66 x 45.7 cm 3/Cs. N/A N/A N/A
Description
Description
  • Large selection of cost-effective, reusable stock containers available
  • Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter
Specifications
Specifications
Insulated Shipper
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.