Learn More
Medchemexpress LLC HY-10201A 500mg , Sorafenib (Tosylate) Bay 43-9006 CAS:475207-59-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10201A500MG
Encompass_Preferred
Medchemexpress, HY-10201A 500mg Sorafenib (Tosylate) Bay 43-9006 CAS:475207-59-1 Formula:C28H24ClF3N4O6S IC50: 6 nM (Raf-1), 20 nM (VEGFR-3), 22 nM (BRAF), 57 nM (PDGFR-β), 58 nM (Flt3), 68 nM (c-KIT), 90 nM (VEGFR-2) Purity:98% Sorafenib tosylate is a potent multikinase inhibitor, with IC 50 s of 6 nM, 20 nM, and 22 nM for Raf-1 , B-Raf , and VEGFR-3 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2351
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.